Gene Transcripts

Gene Symbol
1810010H24Rik
Entrez Gene ID
69066
Gene Name
RIKEN cDNA 1810010H24 gene
Gene Aliases
OTTMUSG00000003581
Chromosome Location
Chr. 11: 107023268 - 107030446 on Build GRCm38
UniGene ID
Mm.40679
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163473.1 NP_001156945.1 3 862
GenBank mRNA AK007422.1 - 3 965
BB826454.1 - 1 191

View Full Product Details