Gene Transcripts

Gene Symbol
9430069I07Rik
Entrez Gene ID
77358
Gene Name
RIKEN cDNA 9430069I07 gene
Gene Aliases
Chromosome Location
Chr. 15: 34349051 - 34357872 on Build GRCm38
UniGene ID
Mm.402759
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256161.1 NP_001243090.1 1 22
GenBank mRNA AK020480.1 - 1 24

View Full Product Details