Gene Transcripts

Gene Symbol
Gdf9
Entrez Gene ID
59304
Gene Name
growth differentiation factor 9
Gene Aliases
Chromosome Location
Chr. 10: 38782499 - 38793238 on Build Rnor_6.0
UniGene ID
Rn.42874
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021672.1 NP_067704.1 2 607
XM_008767704.1 XP_008765926.1 3 524
GenBank mRNA AF099912.1 - 2 607
X81899.1 - 2 607

View Full Product Details