Gene Transcripts

Gene Symbol
Vti1a
Entrez Gene ID
65277
Gene Name
vesicle transport through interaction with t-SNAREs 1A
Gene Aliases
Vti1
Chromosome Location
Chr. 1: 276310072 - 276508635 on Build Rnor_6.0
UniGene ID
Rn.154408
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023101.1 NP_075589.1 3 235
GenBank mRNA AF262221.1 - 3 235
AF262222.1 - 3 221

View Full Product Details