Gene Transcripts

Gene Symbol
Rbbp6
Entrez Gene ID
19647
Gene Name
retinoblastoma binding protein 6
Gene Aliases
4933422O15Rik, AI316869, BB233631, C030034J04Rik, C77662, P2P-R, PACT
Chromosome Location
Chr. 7: 122970546 - 123002572 on Build GRCm38
UniGene ID
Mm.4480
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011247.2 NP_035377.2 10 1507
GenBank mRNA U28789.1 - 4 501
U83913.1 - 5 650

View Full Product Details