Gene Transcripts
- Gene Symbol
- TPMT
- Entrez Gene ID
- 7172
- Gene Name
- thiopurine S-methyltransferase
- Gene Aliases
- TPMTD
- Chromosome Location
- Chr. 6: 18128311 - 18155165 on Build GRCh38
- UniGene ID
- Hs.444319
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000367.3 |
NP_000358.1
|
2, 3 |
311 |
|
XM_011514839.2 |
XP_011513141.1
|
2, 3 |
269 |
|
XM_017011241.1 |
XP_016866730.1
|
3, 4 |
605 |
|
GenBank mRNA |
AF021876.1 |
- |
1, 2 |
134 |
|
AF021877.1 |
- |
1, 2 |
134 |
|
AK310420.1 |
- |
2, 3 |
205 |
|
BC005339.1 |
- |
2, 3 |
221 |
3829338 |
BC009596.1 |
- |
2, 3 |
220 |
4133029 |
BI598325.1 |
- |
2, 3 |
296 |
|
DB481604.1 |
- |
2, 3 |
315 |
|
S62904.1 |
- |
1, 2 |
199 |
|
U12387.1 |
- |
2, 3 |
210 |
|
View Full Product Details