Gene Transcripts

Gene Symbol
1700026D08Rik
Entrez Gene ID
75556
Gene Name
RIKEN cDNA 1700026D08 gene
Gene Aliases
AW047638, AW551802
Chromosome Location
Chr. 7: 83774101 - 83795384 on Build GRCm38
UniGene ID
Mm.17649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029335.3 NP_083611.2 3 280
GenBank mRNA AK006375.1 - 3 268
BC048079.1 - 3 260
BC061016.1 - 3 274 6771412

View Full Product Details