Gene Transcripts

Gene Symbol
Rho
Entrez Gene ID
212541
Gene Name
rhodopsin
Gene Aliases
Noerg1, Opn2, Ops, RP4
Chromosome Location
Chr. 6: 115926754 - 115940036 on Build GRCm38
UniGene ID
Mm.2965
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145383.1 NP_663358.1 3 677
GenBank mRNA AK044333.1 - 3 697
AK044412.1 - 3 695
BC013125.1 - 3 677 4500760
BC031766.1 - 3 678 4527040

View Full Product Details