Gene Transcripts

Gene Symbol
Olfr1233
Entrez Gene ID
258974
Gene Name
olfactory receptor 1233
Gene Aliases
MOR238-1
Chromosome Location
Chr. 2: 89339312 - 89342391 on Build GRCm38
UniGene ID
Mm.223365
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146972.1 NP_667183.1 1 394
GenBank mRNA BC119149.1 - 1 442
BC119151.1 - 1 442 8733900

View Full Product Details