Gene Transcripts

Gene Symbol
Rbbp6
Entrez Gene ID
19647
Gene Name
retinoblastoma binding protein 6
Gene Aliases
4933422O15Rik, AI316869, BB233631, C030034J04Rik, C77662, P2P-R, PACT
Chromosome Location
Chr. 7: 122970546 - 123002572 on Build GRCm38
UniGene ID
Mm.4480
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175023.3 NP_778188.1 3 1788
GenBank mRNA AK045635.1 - 3 1533
AK082686.1 - 1 1130
AK164942.1 - 1 168

View Full Product Details