Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007224.3 | NP_009155.1 | 2 | 1039 | |
XM_017018747.1 | XP_016874236.1 | 2, 3 | 523 | ||
GenBank mRNA | AF043469.1 | - | 1 | 192 | |
AK094231.1 | - | 3 | 1244 | ||
AK290949.1 | - | 2 | 977 | ||
AY358614.1 | - | 2 | 1030 | ||
BC036679.1 | - | 2 | 1033 | 5264190 | |
BC053581.1 | - | 2 | 963 | 5174266 |