Gene Transcripts
- Gene Symbol
- ZNF276
- Entrez Gene ID
- 92822
- Gene Name
- zinc finger protein 276
- Gene Aliases
- CENP-Z, CENPZ, ZADT, ZFP276, ZNF477
- Chromosome Location
- Chr. 16: 89720368 - 89740925 on Build GRCh38
- UniGene ID
- Hs.290154
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113525.1 |
NP_001106997.1
|
5, 6 |
1190 |
|
NM_152287.3 |
NP_689500.2
|
5, 6 |
1180 |
|
NR_110122.1 |
|
5, 6 |
1180 |
|
XM_017023889.1 |
XP_016879378.1
|
5, 6 |
1190 |
|
GenBank mRNA |
AK056405.1 |
- |
5, 6 |
1131 |
|
AK295770.1 |
- |
3, 4 |
852 |
|
AK315793.1 |
- |
2, 3 |
547 |
|
BC032781.1 |
- |
4, 5 |
849 |
|
BC107603.1 |
- |
5, 6 |
1005 |
40036180 |
BC110348.1 |
- |
5, 6 |
905 |
5196957 |
View Full Product Details