Gene Transcripts

Gene Symbol
ZNF273
Entrez Gene ID
10793
Gene Name
zinc finger protein 273
Gene Aliases
HZF9
Chromosome Location
Chr. 7: 64882493 - 64937316 on Build GRCh38
UniGene ID
Hs.520889
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021148.2 NP_066971.2 4 2491
NR_003099.1 5 2580
GenBank mRNA AL832810.1 - 1 2183
BC063818.1 - 5 2575 6503687

View Full Product Details