Gene Transcripts

Gene Symbol
Coil
Entrez Gene ID
12812
Gene Name
coilin
Gene Aliases
C79982, Cln80, p80, p80-coilin
Chromosome Location
Chr. 11: 88973935 - 88991613 on Build GRCm38
UniGene ID
Mm.268004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016706.2 NP_057915.2 4, 5 1492
GenBank mRNA AA240407.1 - 2, 3 92
AF208663.1 - 4, 5 1474
AK146366.1 - 4, 5 1476
BC031167.1 - 4, 5 1507 4983919
CF105200.1 - 3, 4 290

View Full Product Details