Gene Transcripts

Gene Symbol
Coil
Entrez Gene ID
12812
Gene Name
coilin
Gene Aliases
C79982, Cln80, p80, p80-coilin
Chromosome Location
Chr. 11: 88973935 - 88991613 on Build GRCm38
UniGene ID
Mm.268004
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016706.2 NP_057915.2 1 112
GenBank mRNA AF208663.1 - 1 97
AK146366.1 - 1 99
BC031167.1 - 1 130 4983919
BY065340.1 - 1 113

Gene Symbol
Gm10367
Entrez Gene ID
100316846
Gene Name
predicted gene 10367
Gene Aliases
ENSMUSG00000072564
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.415909
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK147092.1 - 1 113

View Full Product Details