Gene Transcripts

Gene Symbol
Ide
Entrez Gene ID
15925
Gene Name
insulin degrading enzyme
Gene Aliases
1300012G03Rik, 4833415K22Rik, AA675336, AI507533
Chromosome Location
Chr. 19: 37268741 - 37334544 on Build GRCm38
UniGene ID
Mm.28366
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031156.3 NP_112419.3 13 1744
GenBank mRNA AJ278422.1 - 13 1764
AK004972.2 - 13 1745
AK128986.1 - 13 1761
BC041675.1 - 13 1776 4951971

View Full Product Details