Gene Transcripts

Gene Symbol
2010002M12Rik
Entrez Gene ID
112419
Gene Name
RIKEN cDNA 2010002M12 gene
Gene Aliases
AW412491, Ifit1b
Chromosome Location
Chr. 19: 34617051 - 34640743 on Build GRCm38
UniGene ID
Mm.440727
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053217.2 NP_444447.1 3 1308
GenBank mRNA AK136525.1 - 2 1160
AK153247.1 - 3 1309
BC138202.1 - 2 1139 8861221

View Full Product Details