Gene Transcripts

Gene Symbol
Ankrd52
Entrez Gene ID
237615
Gene Name
ankyrin repeat domain 52
Gene Aliases
6430544C07, G431002C21Rik
Chromosome Location
Chr. 10: 128377124 - 128394006 on Build GRCm38
UniGene ID
Mm.440177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172790.2 NP_766378.1 21 2326
GenBank mRNA AK090124.1 - 21 2326
BC117907.1 - 21 2277 40097189
BC117908.1 - 21 2277 40097194

View Full Product Details