Gene Transcripts
- Gene Symbol
- Txnl4a
- Entrez Gene ID
- 27366
- Gene Name
- thioredoxin-like 4A
- Gene Aliases
- D18Wsu98e, Dim1, ENSMUSG00000057130, Txnl4, U5-15kD, U5-15kDa
- Chromosome Location
- Chr. 18: 80206794 - 80225851 on Build GRCm38
- UniGene ID
- Mm.172411
- Species
- Mus musculus
- Gene Symbol
- Rp9
- Entrez Gene ID
- 55934
- Gene Name
- retinitis pigmentosa 9 (human)
- Gene Aliases
- PAP-1, Rp9h
- Chromosome Location
- Chr. 9: 22448311 - 22468356 on Build GRCm38
- UniGene ID
- Mm.38235
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC096578.1 |
- |
|
120 |
|
View Full Product Details