Gene Transcripts

Gene Symbol
Acbd3
Entrez Gene ID
289312
Gene Name
acyl-CoA binding domain containing 3
Gene Aliases
Chromosome Location
Chr. 13: 99004942 - 99035520 on Build Rnor_6.0
UniGene ID
Rn.57036
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182843.1 NP_878263.1 8 1514
GenBank mRNA AY336075.1 - 8 1514
BC083877.1 - 8 1475 7130229

View Full Product Details