Gene Transcripts

Gene Symbol
Nfatc2
Entrez Gene ID
311658
Gene Name
nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2
Gene Aliases
Chromosome Location
Chr. 3: 165245238 - 165360292 on Build Rnor_6.0
UniGene ID
Rn.17773
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107805.1 NP_001101275.1 9 2157
XM_008762507.1 XP_008760729.1 9 2151

View Full Product Details