Gene Transcripts

Gene Symbol
Ifnk
Entrez Gene ID
313152
Gene Name
interferon kappa
Gene Aliases
Chromosome Location
Chr. 5: 50631217 - 50634142 on Build Rnor_6.0
UniGene ID
Rn.104672
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107925.1 NP_001101395.1 1 431

View Full Product Details