Gene Transcripts

Gene Symbol
Kif23
Entrez Gene ID
315740
Gene Name
kinesin family member 23
Gene Aliases
Chromosome Location
Chr. 8: 66866086 - 66893179 on Build Rnor_6.0
UniGene ID
Rn.63734
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108155.1 NP_001101625.1 13, 14 1490

View Full Product Details