Gene Transcripts

Gene Symbol
Ugt8
Entrez Gene ID
50555
Gene Name
UDP glycosyltransferase 8
Gene Aliases
Ugt8a
Chromosome Location
Chr. 2: 229644479 - 229718672 on Build Rnor_6.0
UniGene ID
Rn.9744
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019276.3 NP_062149.1 6 1355
GenBank mRNA L21698.1 - 6 1402

View Full Product Details