Gene Transcripts

Gene Symbol
Ahcy
Entrez Gene ID
29443
Gene Name
adenosylhomocysteinase
Gene Aliases
Chromosome Location
Chr. 3: 150587833 - 150603057 on Build Rnor_6.0
UniGene ID
Rn.5878
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017201.1 NP_058897.1 9 1120
GenBank mRNA BC061841.1 - 9 1147
M15185.1 - 9 1120

View Full Product Details