Gene Transcripts

Gene Symbol
Cldn15
Entrez Gene ID
304388
Gene Name
claudin 15
Gene Aliases
Chromosome Location
Chr. 12: 22738198 - 22750410 on Build Rnor_6.0
UniGene ID
Rn.202600
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107135.2 NP_001100605.1 1 173
XM_006249155.2 XP_006249217.1 2 1821

View Full Product Details