Gene Transcripts

Gene Symbol
Rhob
Entrez Gene ID
64373
Gene Name
ras homolog family member B
Gene Aliases
Arhb
Chromosome Location
Chr. 6: 33689127 - 33691301 on Build Rnor_6.0
UniGene ID
Rn.2042
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022542.1 NP_071987.1 1 858
GenBank mRNA M74295.1 - 1 858

View Full Product Details