Gene Transcripts

Gene Symbol
CLK2P1
Entrez Gene ID
1197
Gene Name
CDC like kinase 2, pseudogene 1
Gene Aliases
CLK2P
Chromosome Location
Chr. 7: 23584716 - 23586527 on Build GRCh38
UniGene ID
Hs.632635
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002711.3 1 272
GenBank mRNA BC022025.1 - 1 288


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DB054291.1 - 1 291 Chr. 7 - 23586052 - 23586546 [-]
GenBankmRNA DB036023.1 - 1 259 Chr. 7 - 23585949 - 23586514 [-]
GenBankmRNA DB038453.1 - 1 280 Chr. 7 - 23585969 - 23586534 [-]
GenBankmRNA DB057698.1 - 1 279 Chr. 7 - 23586002 - 23586534 [-]
GenBankmRNA DB446583.1 - 1 269 Chr. 7 - 23586097 - 23586524 [-]
GenBankmRNA HY007936.1 - 1 272 Chr. 7 - 23585933 - 23586527 [-]
GenBankmRNA DB460497.1 - 1 283 Chr. 7 - 23586051 - 23586538 [-]
GenBankmRNA DB023440.1 - 1 315 Chr. 7 - 23586020 - 23586570 [-]
GenBankmRNA DB032385.1 - 1 297 Chr. 7 - 23585968 - 23586552 [-]
GenBankmRNA HY031637.1 - 1 298 Chr. 7 - 23586094 - 23586551 [-]
GenBankmRNA DB022984.1 - 1 284 Chr. 7 - 23586052 - 23586538 [-]
GenBankmRNA BM563567.1 - 261 Chr. - - [-]
GenBankmRNA DB443950.1 - 1 283 Chr. 7 - 23586087 - 23586538 [-]
GenBankmRNA DB056472.1 - 1 283 Chr. 7 - 23586052 - 23586538 [-]
GenBankmRNA HY021239.1 - 1 266 Chr. 7 - 23586048 - 23586520 [-]
GenBankmRNA BG723070.1 - 1 258 Chr. 7 - 23585964 - 23586511 [-]
GenBankmRNA BG717579.1 - 288 Chr. - - [-]
GenBankmRNA BG720946.1 - 1 287 Chr. 7 - 23585791 - 23586538 [-]
GenBankmRNA DB056280.1 - 1 283 Chr. 7 - 23586052 - 23586538 [-]
GenBankmRNA HY032935.1 - 1 255 Chr. 7 - 23586045 - 23586509 [-]
GenBankmRNA HY015400.1 - 1 173 Chr. 7 - 23585892 - 23586428 [-]
GenBankmRNA HY031139.1 - 1 266 Chr. 7 - 23586058 - 23586520 [-]
GenBankmRNA HY005607.1 - 1 266 Chr. 7 - 23586020 - 23586520 [-]
GenBankmRNA DB458507.1 - 1 283 Chr. 7 - 23586061 - 23586538 [-]
GenBankmRNA HY220942.1 - 1 220 Chr. 7 - 23586011 - 23586442 [-]
GenBankmRNA DB032346.1 - 1 276 Chr. 7 - 23586026 - 23586531 [-]
GenBankmRNA DB025100.1 - 1 283 Chr. 7 - 23585991 - 23586538 [-]

View Full Product Details