Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_026785.1 | 1 | 1877 | ||
GenBank mRNA | AK026503.1 | - | 1 | 1787 | |
BC000203.1 | - | 1 | 1790 | ||
BM968345.1 | - | 1 | 455 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM930735.1 | - | 1 | 492 | Chr. 8 - 143366621 - 143367250 [-] |