Gene Transcripts
- Gene Symbol
- ZNF415
- Entrez Gene ID
- 55786
- Gene Name
- zinc finger protein 415
- Gene Aliases
- ZfLp
- Chromosome Location
- Chr. 19: 53107879 - 53133077 on Build GRCh38
- UniGene ID
- Hs.147765
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028343.1 |
|
6, 7 |
632 |
|
XM_006723267.1 |
XP_006723330.1
|
5, 6 |
589 |
|
XM_006723272.2 |
XP_006723335.1
|
4, 5 |
369 |
|
XM_006723273.2 |
XP_006723336.1
|
4, 5 |
453 |
|
XM_006723274.2 |
XP_006723337.1
|
4, 5 |
530 |
|
XM_017026953.1 |
XP_016882442.1
|
5, 6 |
736 |
|
XM_017026954.1 |
XP_016882443.1
|
5, 6 |
613 |
|
XM_017026955.1 |
XP_016882444.1
|
5, 6 |
564 |
|
XM_017026956.1 |
XP_016882445.1
|
6, 7 |
714 |
|
XM_017026957.1 |
XP_016882446.1
|
5, 6 |
452 |
|
XM_017026958.1 |
XP_016882447.1
|
6, 7 |
768 |
|
XM_017026959.1 |
XP_016882448.1
|
5, 6 |
639 |
|
XM_017026960.1 |
XP_016882449.1
|
4, 5 |
354 |
|
XM_017026961.1 |
XP_016882450.1
|
5, 6 |
800 |
|
XM_017026970.1 |
XP_016882459.1
|
5, 6 |
606 |
|
XM_017026971.1 |
XP_016882460.1
|
5, 6 |
825 |
|
XM_017026973.1 |
XP_016882462.1
|
4, 5 |
599 |
|
XM_017026974.1 |
XP_016882463.1
|
5, 6 |
678 |
|
GenBank mRNA |
AK095504.1 |
- |
6, 7 |
604 |
|
DQ925696.1 |
- |
4, 5 |
442 |
|
DQ925697.1 |
- |
5, 6 |
452 |
|
View Full Product Details