Gene Transcripts

Gene Symbol
TNS3
Entrez Gene ID
64759
Gene Name
tensin 3
Gene Aliases
TEM6, TENS1
Chromosome Location
Chr. 7: 47275154 - 47582553 on Build GRCh38
UniGene ID
Hs.520814
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011515484.2 XP_011513786.1 1 330
GenBank mRNA AL137468.1 - 1 278

View Full Product Details