Gene Transcripts
- Gene Symbol
- Gm15179
- Entrez Gene ID
- 100503248
- Gene Name
- predicted gene 15179
- Gene Aliases
- OTTMUSG00000019417
- Chromosome Location
- Chr. 1: 75368210 - 75375015 on Build GRCm38
- UniGene ID
- Mm.403916
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037976.1 |
|
4 |
644 |
|
GenBank mRNA |
AK039780.1 |
- |
4 |
642 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB198141.2 |
- |
4 |
650 |
|
Chr. 1 - 75368212 - 75374992 [-] |
View Full Product Details