Gene Transcripts
- Gene Symbol
- Ern1
- Entrez Gene ID
- 78943
- Gene Name
- endoplasmic reticulum (ER) to nucleus signalling 1
- Gene Aliases
- 9030414B18Rik, AI225830, C85377, Ire1a, Ire1alpha, Ire1p
- Chromosome Location
- Chr. 11: 106397620 - 106487796 on Build GRCm38
- UniGene ID
- Mm.340943
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK083891.1 |
- |
1 |
1328 |
|
View Full Product Details