Gene Transcripts
- Gene Symbol
- 8030443L12Rik
- Entrez Gene ID
- 77546
- Gene Name
- RIKEN cDNA 8030443L12 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK020200.1 |
- |
1 |
376 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB749871.1 |
- |
1 |
352 |
|
Chr. 1 - 131084274 - 131084711 [-] |
GenBankmRNA |
BY720421.1 |
- |
1 |
371 |
|
Chr. 1 - 131084262 - 131084731 [-] |
View Full Product Details