Gene Transcripts
- Gene Symbol
- Gm30146
- Entrez Gene ID
- 102631944
- Gene Name
- predicted gene, 30146
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK082843.1 |
- |
3 |
594 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AV586744.1 |
- |
2 |
455 |
|
Chr. 7 - 25860203 - 25869894 [+] |
GenBankmRNA |
BY728877.1 |
- |
3 |
594 |
|
Chr. 7 - 25860140 - 25869883 [+] |
View Full Product Details