Gene Transcripts
- Gene Symbol
- Gm26787
- Entrez Gene ID
- 102637672
- Gene Name
- predicted gene, 26787
- Gene Aliases
- Chromosome Location
- Chr. 9: 36767767 - 36782191 on Build GRCm38
- UniGene ID
- Mm.398336
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007186.1 |
- |
4 |
515 |
|
AK018988.1 |
- |
4 |
494 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707377.1 |
- |
4 |
494 |
|
Chr. 9 - 36767910 - 36782677 [+] |
View Full Product Details