Gene Transcripts
- Gene Symbol
- Csnk1d
- Entrez Gene ID
- 104318
- Gene Name
- casein kinase 1, delta
- Gene Aliases
- 1200006A05Rik, AA409348, D930010H05Rik
- Chromosome Location
- Chr. 11: 120958010 - 120991333 on Build GRCm38
- UniGene ID
- Mm.216227
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK042000.1 |
- |
1 |
3027 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB234840.2 |
- |
1 |
249 |
|
Chr. 11 - 120975965 - 120976536 [-] |
View Full Product Details