Gene Transcripts
- Gene Symbol
- 4933415J04Rik
- Entrez Gene ID
- 71172
- Gene Name
- RIKEN cDNA 4933415J04 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.79094
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016822.1 |
- |
1 |
766 |
|
View Full Product Details