Gene Transcripts
- Gene Symbol
- LOC100505664
- Entrez Gene ID
- 100505664
- Gene Name
- uncharacterized LOC100505664
- Gene Aliases
- Chromosome Location
- Chr. 20: 23159640 - 23190307 on Build GRCh38
- UniGene ID
- Hs.120058
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091765.1 |
- |
1 |
2186 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000411595.2 |
- |
|
553 |
|
Chr. 20 - 23188128 - 23190307 [+] |
View Full Product Details