Gene Transcripts
- Gene Symbol
- IQCF5-AS1
- Entrez Gene ID
- 101928999
- Gene Name
- IQCF5 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 3: 51873596 - 51875767 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109984.1 |
|
3 |
642 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000440723.1 |
- |
3 |
642 |
|
Chr. 3 - 51873596 - 51875767 [+] |
View Full Product Details