Gene Transcripts
- Gene Symbol
- LINC01197
- Entrez Gene ID
- 400456
- Gene Name
- long intergenic non-protein coding RNA 1197
- Gene Aliases
- Chromosome Location
- Chr. 15: 95279288 - 95327100 on Build GRCh38
- UniGene ID
- Hs.536404
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_034095.1 |
|
3 |
442 |
|
GenBank mRNA |
BC040875.1 |
- |
3 |
442 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000508732.6 |
- |
3 |
442 |
|
Chr. 15 - 95279284 - 95327100 [-] |
View Full Product Details