Gene Transcripts
- Gene Symbol
- LOC102724064
- Entrez Gene ID
- 102724064
- Gene Name
- uncharacterized LOC102724064
- Gene Aliases
- Chromosome Location
- Chr. 11: 66347991 - 66363764 on Build GRCh38
- UniGene ID
- Hs.614641
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_135761.1 |
|
1, 2 |
89 |
|
GenBank mRNA |
BM727141.1 |
- |
1, 2 |
89 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM719798.1 |
- |
1, 2 |
89 |
|
Chr. 11 - 66347991 - 66363730 [+] |
GenBankmRNA |
BM727596.1 |
- |
1, 2 |
89 |
|
Chr. 11 - 66347991 - 66363451 [+] |
View Full Product Details