Gene Transcripts
- Gene Symbol
- Gm11482
- Entrez Gene ID
- 102632940
- Gene Name
- predicted gene 11482
- Gene Aliases
- OTTMUSG00000001234
- Chromosome Location
- Chr. 4: 63634415 - 63641981 on Build GRCm38
- UniGene ID
- Mm.399154
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK077143.1 |
- |
3 |
252 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BU939496.1 |
- |
|
221 |
|
Chr. - - [-] |
GenBankmRNA |
BB617036.1 |
- |
3 |
251 |
|
Chr. 4 - 63634421 - 63639361 [+] |
View Full Product Details