Gene Transcripts
- Gene Symbol
- SH3PXD2A-AS1
- Entrez Gene ID
- 100505839
- Gene Name
- SH3PXD2A antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 103746779 - 103755409 on Build GRCh38
- UniGene ID
- Hs.655149
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038940.1 |
|
1, 2 |
311 |
|
GenBank mRNA |
AK056784.1 |
- |
1, 2 |
93 |
|
DA422325.1 |
- |
1, 2 |
311 |
|
View Full Product Details