Gene Transcripts

Gene Symbol
Vmn1r42
Entrez Gene ID
113848
Gene Name
vomeronasal 1 receptor 42
Gene Aliases
V1ra1, V1ra3, V1ra6
Chromosome Location
Chr. 6: 89844518 - 89845615 on Build GRCm38
UniGene ID
Mm.349358
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053221.2 NP_444451.2 1 901
GenBank mRNA BC125372.1 - 1 901

View Full Product Details