Gene Transcripts

Gene Symbol
Gm7073
Entrez Gene ID
631784
Gene Name
predicted gene 7073
Gene Aliases
Chromosome Location
Chr. X: 60436052 - 60456195 on Build GRCm38
UniGene ID
Mm.424878
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039240.3 NP_001034329.2 4 972
GenBank mRNA AK135660.1 - 4 974

View Full Product Details