Gene Transcripts

Gene Symbol
Timm8b
Entrez Gene ID
64372
Gene Name
translocase of inner mitochondrial membrane 8 homolog b (yeast)
Gene Aliases
Ddp2
Chromosome Location
Chr. 8: 55037750 - 55039130 on Build Rnor_6.0
UniGene ID
Rn.3792
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022541.2 NP_071986.1 1, 2 105
GenBank mRNA AF196315.1 - 1, 2 106
CF977529.1 - 1, 2 115

View Full Product Details