Gene Transcripts
- Gene Symbol
- Rhob
- Entrez Gene ID
- 64373
- Gene Name
- ras homolog family member B
- Gene Aliases
- Arhb
- Chromosome Location
- Chr. 6: 33689127 - 33691301 on Build Rnor_6.0
- UniGene ID
- Rn.2042
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022542.1 |
NP_071987.1
|
1 |
398 |
|
GenBank mRNA |
M74295.1 |
- |
1 |
398 |
|
View Full Product Details