Gene Transcripts

Gene Symbol
Chic2
Entrez Gene ID
83835
Gene Name
cysteine-rich hydrophobic domain 2
Gene Aliases
Chromosome Location
Chr. 14: 35683657 - 35718005 on Build Rnor_6.0
UniGene ID
Rn.106037
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105736.2 NP_001099206.1 4 403
GenBank mRNA BE109715.1 - 5 381
BF558018.1 - 2 181
EU202679.1 - 4 349
EU202680.1 - 4 349

View Full Product Details