Gene Transcripts

Gene Symbol
Tubb3
Entrez Gene ID
246118
Gene Name
tubulin, beta 3 class III
Gene Aliases
Chromosome Location
Chr. 19: 56220759 - 56229813 on Build Rnor_6.0
UniGene ID
Rn.43958
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139254.2 NP_640347.2 2 194
GenBank mRNA BC097281.1 - 2 174
CF976541.1 - 2 198

View Full Product Details